4-Fluorsalicylaldehyd
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS31258G5EA
0
CHF
ACTIAS31258G5
ACTIAS31258G1
4-Fluorsalicylaldehyd
4-Fluorsalicylaldehyd
Formel:
C₇H₅FO₂ MW: 140,1118 g/mol |
MDL Number:
MFCD03788526 CAS-Nummer: 348-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...