4-Brom-1-naphthol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS30852G5EA 0 CHF
ACTIAS30852G5 ACTIAS30852G1
4-Brom-1-naphthol
4-Brom-1-naphthol
Formel: C₁₀H₇BrO
MW: 223,0700 g/mol
MDL Number: MFCD03129265
CAS-Nummer: 571-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR