4-Methyl-3-(trifluoromethyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32331G5EA 0 CHF
ACTIAS32331G5 ACTIAS32331G10
4-Methyl-3-(trifluoromethyl)benzoic acid
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MDL Number: MFCD01631592
CAS Number: 261952-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR