4-Bromo-3-(trifluoromethyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40401G5EA 0 CHF
ACTIAS40401G5
4-Bromo-3-(trifluoromethyl)benzoic acid
4-Bromo-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
MDL Number: MFCD08459288
CAS Number: 161622-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR