3-Bromo-p-anisidine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3-Bromo-4-methoxyaniline
ACTIAS35076G5EA
0
CHF
ACTIAS35076G10
ACTIAS35076G1
ACTIAS35076G5
3-Bromo-p-anisidine
3-Bromo-p-anisidine
Formula:
C₇H₈BrNO |
MDL Number:
MFCD04094339 CAS Number: 19056-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...