4-Isopropylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Isopropylbenzeneboronic acid

ACTIAS50852G100EA 0 CHF
ACTIAS50852G100 ACTIAS50852G15
4-Isopropylphenylboronic acid
4-Isopropylphenylboronic acid
Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
MDL Number: MFCD01074614
CAS Number: 16152-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR