Rhodium(II)acetat dimer

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS40695G1EA 0 CHF
ACTIAS40695G1
Rhodium(II)acetat dimer
Rhodium(II)acetat dimer
Formel: C₈H₁₂O₈Rh₂
MW: 441,987 g/mol
MDL Number: MFCD00003538
CAS-Nummer: 15956-28-2
EINECS: 240-084-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR