5-Nitro-2-thiophenecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31519G5EA 0 CHF
ACTIAS31519G5 ACTIAS31519G1
5-Nitro-2-thiophenecarboxylic acid
5-Nitro-2-thiophenecarboxylic acid
Formula: C₅H₃NO₄S
MW: 173.14
MDL Number: MFCD00159552
CAS Number: 6317-37-9
EINECS: 228-654-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR