Cyclopropylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50178G5EA 0 CHF
ACTIAS50178G5 ACTIAS50178G25 ACTIAS50178G1
Cyclopropylboronic acid
Cyclopropylboronic acid
Formula: C₃H₇BO₂
MDL Number: MFCD04038750
CAS Number: 411235-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR