Cyclopropylboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

ACTIAS33294G5EA 0 CHF
ACTIAS33294G5 ACTIAS33294G1
Cyclopropylboronic acid pinacol ester
Cyclopropylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
MW: 168.04
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR