Benzo[b]thiophen-3-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31492G5EA 0 CHF
ACTIAS31492G5
Benzo[b]thiophen-3-carbaldehyd
Benzo[b]thiophen-3-carbaldehyd
Formel: C₉H₆OS
MW: 162,2083 g/mol
MDL Number: MFCD00052376
CAS-Nummer: 5381-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR