Benzo[b]thiophen-2-carbaldehyd
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS32740G1EA
0
CHF
ACTIAS32740G1
ACTIAS32740G10
ACTIAS32740G5
Benzo[b]thiophen-2-carbaldehyd
Benzo[b]thiophen-2-carbaldehyd
Formel:
C₉H₆OS MW: 162,21 g/mol |
MDL Number:
MFCD01075041 CAS-Nummer: 3541-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...