3-(Hydroxymethyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(Hydroxymethyl)benzeneboronic acid

ACTIAS50819G5EA 0 CHF
ACTIAS50819G5 ACTIAS50819G25 ACTIAS50819G1
3-(Hydroxymethyl)phenylboronic acid
3-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD01317846
CAS Number: 87199-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR