2-(Hydroxymethyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-(Hydroxymethyl)benzeneboronic acid

ACTIAS51456G5EA 0 CHF
ACTIAS51456G5 ACTIAS51456G1 ACTIAS51456G25
2-(Hydroxymethyl)phenylboronic acid
2-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR