Methyl-4-amino-3-hydroxybenzoat
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS36575G1EA
0
CHF
ACTIAS36575MG250
ACTIAS36575G1
ACTIAS36575G5
Methyl-4-amino-3-hydroxybenzoat
Methyl-4-amino-3-hydroxybenzoat
Formel:
C₈H₉NO₃ MW: 167,17 g/mol |
MDL Number:
MFCD00017093 CAS-Nummer: 63435-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...