Methyl-4-aminosalicylat

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS21741G25EA 25 CHF
ACTIAS21741G25 ACTIAS21741G5
Methyl-4-aminosalicylat
Methyl-4-aminosalicylat
Formel: C₈H₉NO₃
MW: 167,18 g/mol
MDL Number: MFCD00088091
CAS-Nummer: 4136-97-4
EINECS: 223-958-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR