Potassium 2-phenylethyltrifluoroborate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Potassium Trifluoro(2-phenylethyl)boratePotassium PhenethyltrifluoroboratePotassium Trifluoro(phenethyl)borate

ACTIAS24461G5EA 247 CHF
ACTIAS24461G5
Potassium 2-phenylethyltrifluoroborate
Potassium 2-phenylethyltrifluoroborate
Formula: C₈H₉BF₃K
MW: 212.06 g/mol
MDL Number: MFCD09039257
CAS Number: 329976-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR