Methyl glycolate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Methyl 2-hydroxyacetateGlycolic Acid Methyl EsterHydroxyacetic Acid Methyl EsterMethyl Hydroxyacetate

ACTIAS20654G100EA 65 CHF
ACTIAS20654G100 ACTIAS20654G25
Methyl glycolate
Methyl glycolate
Formula: C₃H₆O₃
MW: 90.08 g/mol
MDL Number: MFCD00004667
CAS Number: 96-35-5
EINECS: 202-502-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR