3-Bromo-1H-1,2,4-triazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40310G5EA 0 CHF
ACTIAS40310G1 ACTIAS40310G5
3-Bromo-1H-1,2,4-triazole
3-Bromo-1H-1,2,4-triazole
Formula: C₂H₂BrN₃
MDL Number: MFCD01421960
CAS Number: 7343-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR