3,5-Dibromo-1H-1,2,4-triazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,5-Dibromo-1,2,4-triazole

ACTIAS31747G1EA 0 CHF
ACTIAS31747G1 ACTIAS31747G5
3,5-Dibromo-1H-1,2,4-triazole
3,5-Dibromo-1H-1,2,4-triazole
Formula: C₂HBr₂N₃
MDL Number: MFCD00487260
CAS Number: 7411-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR