2-Hydroxythiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS36506G5EA 0 CHF
ACTIAS36506G5 ACTIAS36506G1 ACTIAS36506MG250
2-Hydroxythiazole
2-Hydroxythiazole
Formula: C₃H₃NOS
MW: 101.1270 g/mol
MDL Number: MFCD02094166
CAS Number: 82358-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR