2-Amino-4-brom-6-methylpyrimidin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS31034G5EA
0
CHF
ACTIAS31034G5
ACTIAS31034G1
2-Amino-4-brom-6-methylpyrimidin
2-Amino-4-bromo-6-methylpyrimidine
Formel:
C₅H₆BrN₃ |
MDL Number:
MFCD11109799 CAS-Nummer: 5734-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...