2,6-Dichloropyrimidine-4-carbonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS37421G1EA 0 CHF
ACTIAS37421MG250 ACTIAS37421G1
2,6-Dichloropyrimidine-4-carbonitrile
2,6-Dichloropyrimidine-4-carbonitrile
Formula: C₅HCl₂N₃
CAS Number: 26293-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR