1-Phenyl-1H-pyrazol-5-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS33572G1EA 0 CHF
ACTIAS33572G1
1-Phenyl-1H-pyrazol-5-carbaldehyd
1-Phenyl-1H-pyrazol-5-carbaldehyd
Formel: C₁₀H₈N₂O
MDL Number: MFCD00103497
CAS-Nummer: 132274-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR