3-Amino-1H-pyrazole-4-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50561G1EA 0 CHF
ACTIAS50561G1 ACTIAS50561G5
3-Amino-1H-pyrazole-4-carboxylic acid
3-Amino-1H-pyrazole-4-carboxylic acid
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
MDL Number: MFCD00005239
CAS Number: 41680-34-6
EINECS: 255-493-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR