Hand cleansing creams, Solopol® Light
Supplier: SC Johnson
Solopol® Light
115-1497EA 0 CHF
115-1497
Hand cleansing creams, Solopol® Light
Skincare Soaps and Skin Cleansers
Solvent-free hand cleanser paste with scrubbing agent for medium- to heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust or lubricants.
  • With extremely fine granulated Astopon® (refined walnut shell powder)
  • Contains skin protectant Eucornol® and aloe vera
  • Effective cleansing with conditioned skin feel
  • Non soapy, solvent-free, colorant-free, perfumed
Certifications: EU Ecolabel
Ordering information: Soft bottle may only be used with Stoko® Vario® dispensers.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904