2-Fluor-6-methoxybenzonitril
Lieferant: Apollo Scientific
APOSPC3817F-100GEA
73.3
CHF
APOSPC3817F-100G
2-Fluor-6-methoxybenzonitril
2-Fluor-6-methoxybenzonitril
Formel:
C₈H₆FNO MW: 151,14 g/mol Siedepunkt: 245 °C (1013 hPa) Melting Pt: 93…95 °C |
MDL Number:
MFCD00042291 CAS-Nummer: 94088-46-7 EINECS: 302-047-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...