Nitric acid 5 mol/l (5 N)

Danger

PRLS85022.400EA 0 CHF
PRLS85022.400
Nitric acid 5 mol/l (5 N)
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR