4-Chloro-2-(trifluoromethoxy)aniline ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine
H34133.MDEA 71.9 CHF
H34133.MD H34133.03
4-Chloro-2-(trifluoromethoxy)aniline ≥97%
4-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
Boiling Pt: 82 °C (15 mmHg)
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00153102
CAS Number: 175205-77-3
UN: 2811
ADR: 6.1,III