2-(3-Methylphenyl)thioacetamide ≥97%
Supplier: Thermo Scientific
Danger
H33031.03EA 122 CHF
H33031.03 H33031.06
2-(3-Methylphenyl)thioacetamide ≥97%
2-(3-Methylphenyl)thioacetamide
Formula: C₉H₁₁NS
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09757580
CAS Number: 6487-91-8