Di-n-octyl phthalate-[ring-D₄] (98% D)
Supplier: LGC Standards PROMOCHEM
Synonyms: Di-n-octyl phthalate-[ring-²H₄]
PROOCILDLM16300.1EA 945 CHF
PROOCILDLM16300.1 CAMBDLM-1630-0.1
Di-n-octyl phthalate-[ring-D₄] (98% D)
Di-n-octyl phthalate-[ring-D4]
Formula: C₂₄H₃₄D₄O₄
MDL Number: MFCD00144150
CAS Number: 93952-13-7
UN: 3082
ADR: 9,III