Iodine 0.4 mol/l (0.8 N) in toluene

PRLSMC306747.0500EA 0 CHF
PRLSMC306747.0500
Iodine 0.4 mol/l (0.8 N) in toluene
Iodine
Formula: I₂
MW: 253.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR