4-Chloro-N-(4-fluorobenzyl)aniline ≥97%
Supplier: Alfa Aesar
Warning
H56981.03EA 397 CHF
H56981.03 H56981.MD
4-Chloro-N-(4-fluorobenzyl)aniline ≥97%
4-Chloro-N-(4-fluorobenzyl)aniline
Formula: C₁₃H₁₁ClFN
MW: 235,69
Storage Temperature: Ambient
MDL Number: MFCD03210761
CAS Number: 356531-46-9
EINECS: 000-000-0
UN: 0000