Diheptyl phthalate (mixture of branched chain isomers)
Supplier: TCI EUROPE
TCIAP0468-25MLEA 0 CHF
TCIAP0468-25ML
Diheptyl phthalate (mixture of branched chain isomers)
Diheptyl phthalate (mixture of branched chain isomers)
Formula: C₂₂H₃₄O₄
MDL Number: MFCD00043680
CAS Number: 41451-28-9