Estriol
CHMCCBL96EA
0
CHF
CHMCCBL96
Estriol
Estriol
Formel:
C₁₈H₂₄O₃ |
MDL Number:
MFCD00003691 CAS-Nummer: 50-27-1 EINECS: 200-022-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...