Single element standards for ICP-OES/MP-AES
Supplier: Agilent Technologies
VARI190026449EA 0 CHF
VARI190026449 VARI190026437 VARI190026415 VARI190026426 VARI190026414 VARI190026425 VARI190026436 VARI190026435 VARI190026424 VARI190026419 VARI190026429 VARI190026418 VARI190026417 VARI190026439 VARI190026441 VARI190026430 VARI190026440 VARI190026423 VARI190026433 VARI190026432 VARI190026421 VARI190026431 VARI190026420
Single element standards for ICP-OES/MP-AES
Solutions are manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
  • Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal
  • Certificates of analysis supplied