Weiter angemeldet bleiben?
Abmeldung in: 
Sie wurden abgemeldet
Möchten Sie sich wieder anmelden?
The VWR Thermocycler Family
The VWR Thermocycler Family
The VWR® Collection Thermal Cycler family
Innovative technology in all formats!
The latest generation of the VWR thermal cycler family, packed with pioneering cycler engineering and now available in 32 well (Ristretto), 2 x 48 well (Doppio), 96 well and 384 well (UNO) block formats.
The combination of high-specification engineering and superior system control enables powerful, fast ramping, stable temperature control and a high level of system reliability. The 'touch screen' control, USB ports, LAN connection and universal block for 0.2 ml and 0.5 ml tubes result in unique ease of use.
The VWR® Collection thermal cyclers provide both best technical specifications and innovative features, e.g. FlexGradient Technology, Emulation Mode, 'PCR Cycler Master' PC software, etc.
Product Video
Watch our product video to get a quick impression of our easy-to-use Thermal cyclers and some of the innovative features like FlexGradient Technology and Emulation Mode.
Product Overview
Ristretto
32 well
Doppio
(gradient optional)
2 x 48 well
UNO96
(gradient optional)
96 well
UNO96 HPL
(gradient optional)
96 well
UNO384 HPL
384 well