Congélateurs
Toutes formes, tailles et températures, avec la même sécurité et la même précision.
Featured Products
VWR Ultra-Low Temperature Upright Freezers
Great value for money – Designed for sample protection and ease of use. Choose from four -86 °C models or two -40 °C models. Four inner doors reduce cold air loss and improve temperature recovery after door openings.
    Learn more
    Resources
    Thermo Scientific Ultra-Low Temperature Upright Freezers, TSX series
    We offer a total package of high quality products within cryopreservation e.g. ULT freezers, racks, cryo boxes, cryotubes and support products like dataloggers and thermometers.
      Learn more
      Chest ULT Freezer
      Great value for money – Designed for sample protection and ease of use. Choose from three -86 °C models. Polystyrene interior sub lids reduce cold air loss and improve temperature recovery after door openings. 
        Learn more
        Resources
        Considerations before buying a freezer
        • What kind of samples do you need to stock?
        • At which temperature do you need to store your samples?
        • How many samples do you need to stock?
        • Do you prefer an upright freezer or a chest freezer?
        • What capacity do you need?
        • Is energy saving most important or is it purchase price?
        • Are additional accessories required, back up system, chart recorder etc.?
        • Do you need to document the temperature? Datalogging?
        • Do you need racks & boxes?
        • Do you need support for the installation? Do you need IQ/OQ/PQ?
          Request a Quote
          Total Freezer Selection
          Our full range of freezers...
          Freezers
          Technical Services
          We help you to keep your laboratory in compliance and in operation
          VWR’s extensive services departments and network of ISO 9001 / IEC17025 accredited service providers, allows us to be your single point of contact for all service needs. For our complete ULT freezer range we can offer:
          • Installation: After a purchase, or after moving to another site, we can install your devices and do all neccessary calibration, qualification and certification steps.
          • Qualification and validation (IQ/OQ/PQ): Qualify your equipment and validate your systems/processes to establish and maintain compliance.
          • Repair: Professional and certified repair.
          • Service contracts: For preventative maintenance, full service and extended warranty
          • Equipment management: our state-of-the-art, cloud-based VEM software helps you to manage all your equipment and instruments. No need to think about due dates for maintenance or manually process service orders.
          Interested? Go to vwr.com/eis to learn more about our equipment and instrument services or contact: services@avantorsciences.com.