X-ray fluorescence spectroscopy (XRF)
Avantor X-ray fluorescence spectroscopy (XRF)
Avantor can provide you with the products and materials you need for to conduct X-ray fluorescence spectroscopy, or XRF, in your lab. Choose the products that work best for you from among our extensive selection.
X-ray fluorescence spectroscopy, or XRF, lets analysts obtain qualitative and quantitative information by measuring the fluorescent X-rays that are emitted from a sample when X-rays are projected onto the sample from an outside source. You can get the products and materials you need to for XRF spectroscopy from a single, trusted source.
XRF Fluorescence Standards
Avantor offers fluorescence standards and solutions in several configurations and formats so you can choose what works best for your laboratory.
XRF Consumables
You can get all the XRF consumables you need, including XRF flux, sample cups, and XRF thin film from Avantor. We carry a wide assortment of consumable products so you can get what you need from brands you trust.